missing translation for 'onlineSavingsMsg'
Learn More
Thallium(III) nitrate trihydrate, 99.5% (metals basis), Thermo Scientific Chemicals Product Code.: 11399139

Thallium(III) nitrate trihydrate, 99.5% (metals basis), Thermo Scientific Chemicals

Product Code. 11399139
5 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11399139

Brand: Thermo Scientific Alfa Aesar 087999.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thallium(III) nitrate trihydrate is a reagent used for the rapid oxidation of styrene derivatives to carbonyl compounds, for the regioselective ring expansion of cyclic aralkyl ketones via oxidative rearrangement, for the selective hydrolysis of thioacetals to carbonyl compounds and for a general synthetic method to 19-norsteroids. Mediates ring contraction of cyclic ketones to a series of trans-fused 10-methyl-2-decalones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13453-38-8
Molecular Formula H12N3O15Tl
Molecular Weight (g/mol) 498.48
MDL Number MFCD00149874
InChI Key TWTZVACKNJEUQF-UHFFFAOYSA-N
Synonym nitric acid,thallium 3+ salt, trihydrate 9ci, thallium trinitrate trihydrate, thallium iii nitrate trihydrate
PubChem CID 11270721
SMILES O.O.O.O.O.O.[Tl+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 102°C to 105°C
Physical Form Crystalline
Quantity 5 g
UN Number UN2727
Solubility Information Soluble in water. (95g/L at 20°C).
IUPAC Name thallium(3+);trinitrate;trihydrate
Formula Weight 444.43 (390.38 Anhydrous)
Percent Purity 99.5%
Odor Odorless
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Thallium (III) nitrate trihydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM03D028 Certificate TypeCertificate of Analysis Date15/01/2025 Catalog Number087999.06,087999.14,087999.22
Lot NumberZ06I030 Certificate TypeCertificate of Analysis Date05/01/2023 Catalog Number087999.06,087999.14,087999.22
Lot NumberL18M22 Certificate TypeCertificate of Analysis Date28/12/2021 Catalog Number087999.06,087999.14,087999.22
Lot NumberG11R023 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number087999.06,087999.14,087999.22
Lot NumberR05G019 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number087999.06,087999.14,087999.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thallium(III) nitrate trihydrate, 99.5% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.