missing translation for 'onlineSavingsMsg'
Learn More

Thallium(III) oxide, 96%, Thermo Scientific Chemicals

57.89 € - 180.90 €

Chemical Identifiers

CAS 1314-32-5
Molecular Formula O3Tl2
Molecular Weight (g/mol) 456.757
MDL Number MFCD00011276
InChI Key QTQRFJQXXUPYDI-UHFFFAOYSA-N
Synonym oxo oxothallanyloxy thallane, tl2o3, thallium iii oxide, oxo oxothallanyl oxy thallane, thallium oxide ic, thallium iii oxide trace metals basis
PubChem CID 3579754
IUPAC Name oxo(oxothallanyloxy)thallane
SMILES O=[Tl]O[Tl]=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11385109
View Documents
Thermo Scientific Alfa Aesar
A17601.09
10 g
57.89 €
10g
Please to purchase this item. Need a web account? Register with us today!
11395109
View Documents
Thermo Scientific Alfa Aesar
A17601.18
50 g
180.90 €
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Thallium(III) oxide is used in organic synthesis and as an analytical reagent

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thallium(III) oxide is used in organic synthesis and as an analytical reagent

Solubility
Insoluble in water and alkalis. Decomposes in HCl and H2SO4

Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications

Chemical Identifiers

1314-32-5
456.757
QTQRFJQXXUPYDI-UHFFFAOYSA-N
3579754
O=[Tl]O[Tl]=O
O3Tl2
MFCD00011276
oxo oxothallanyloxy thallane, tl2o3, thallium iii oxide, oxo oxothallanyl oxy thallane, thallium oxide ic, thallium iii oxide trace metals basis
oxo(oxothallanyloxy)thallane

Specifications

875°C
1314-32-5
MFCD00011276
14,9268
Insoluble in water and alkalis. Decomposes in HCl and H2SO4
O=[Tl]O[Tl]=O
456.757
456.74
Odorless
Thallium(III) oxide
717°C
O3Tl2
UN1707
oxo oxothallanyloxy thallane, tl2o3, thallium iii oxide, oxo oxothallanyl oxy thallane, thallium oxide ic, thallium iii oxide trace metals basis
QTQRFJQXXUPYDI-UHFFFAOYSA-N
oxo(oxothallanyloxy)thallane
3579754
96%
9.65
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H330-H373
Fatal if swallowed.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.

P260-P264b-P270-P271-P284-P301+P310-P304+P340-P310-P314-P330-P501c

H300+H330-H373

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: THALLIUM COMPOUNDS, N.O.S.

missing translation for 'einecsNumber' : 215-229-3

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.