missing translation for 'onlineSavingsMsg'
Learn More

Thallium(III) trifluoroacetate, 95%, Thermo Scientific Chemicals

42.61 € - 498.00 €

Chemical Identifiers

CAS 23586-53-0
Molecular Formula C6F9O6Tl
Molecular Weight (g/mol) 543.426
MDL Number MFCD00000414
InChI Key PSHNNUKOUQCMSG-UHFFFAOYSA-K
Synonym thallium iii trifluoroacetate, acetic acid, trifluoro-, thallium 3+ salt, acetic acid,2,2,2-trifluoro-, thallium 3+ salt 3:1, thallium 3+ ; 2,2,2-trifluoroacetate, thallium tris trifluoroacetate, trifluoroacetic acid thallium iii, thallic trifluoroacetate, tech, thallium 3+ tritrifluoroacetate
PubChem CID 90200
IUPAC Name thallium(3+);2,2,2-trifluoroacetate
SMILES C(=O)(C(F)(F)F)[O-].C(=O)(C(F)(F)F)[O-].C(=O)(C(F)(F)F)[O-].[Tl+3]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11359129
View Documents
Thermo Scientific Alfa Aesar
087913.04
2 g
42.61 €
2g
Please to purchase this item. Need a web account? Register with us today!
11379129
View Documents
Thermo Scientific Alfa Aesar
087913.09
10 g
136.00 €
10g
Please to purchase this item. Need a web account? Register with us today!
11369129
View Documents
Thermo Scientific Alfa Aesar
087913.18
50 g
498.00 €
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Thallium(III) trifluoroacetate is an oxidizing agent used in the synthesis of S-substituted cysteine peptides and proteins containing disulfide bonds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

23586-53-0
543.426
PSHNNUKOUQCMSG-UHFFFAOYSA-K
90200
C(=O)(C(F)(F)F)[O-].C(=O)(C(F)(F)F)[O-].C(=O)(C(F)(F)F)[O-].[Tl+3]
C6F9O6Tl
MFCD00000414
thallium iii trifluoroacetate, acetic acid, trifluoro-, thallium 3+ salt, acetic acid,2,2,2-trifluoro-, thallium 3+ salt 3:1, thallium 3+ ; 2,2,2-trifluoroacetate, thallium tris trifluoroacetate, trifluoroacetic acid thallium iii, thallic trifluoroacetate, tech, thallium 3+ tritrifluoroacetate
thallium(3+);2,2,2-trifluoroacetate

Specifications

213°C (decomposition)
Solid
MFCD00000414
thallium iii trifluoroacetate, acetic acid, trifluoro-, thallium 3+ salt, acetic acid,2,2,2-trifluoro-, thallium 3+ salt 3:1, thallium 3+ ; 2,2,2-trifluoroacetate, thallium tris trifluoroacetate, trifluoroacetic acid thallium iii, thallic trifluoroacetate, tech, thallium 3+ tritrifluoroacetate
PSHNNUKOUQCMSG-UHFFFAOYSA-K
thallium(3+);2,2,2-trifluoroacetate
90200
95%
Thallium (III) trifluoroacetate
23586-53-0
C6F9O6Tl
UN1707
Soluble in water (decomposes).
C(=O)(C(F)(F)F)[O-].C(=O)(C(F)(F)F)[O-].C(=O)(C(F)(F)F)[O-].[Tl+3]
543.426
543.42
Hygroscopic
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H330-H373
Fatal if swallowed.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.

P260-P264b-P270-P271-P284-P301+P310-P304+P340-P310-P314-P330-P501c

H300+H330-H373

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: THALLIUM COMPOUNDS, N.O.S.

missing translation for 'einecsNumber' : 245-761-1

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.