missing translation for 'onlineSavingsMsg'
Learn More
Thallium rod, 12.7mm (0.5in) dia, 99.999% (metals basis), Thermo Scientific Chemicals Product Code.: 11337517

Thallium rod, 12.7mm (0.5in) dia, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 11337517
75 mm
missing translation for 'orderingAttributeHoverText'
Quantity:
75 mm
150 mm
This item is not returnable. View return policy

Product Code. 11337517

Brand: Thermo Scientific Alfa Aesar 012714.0Y

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-28-0
Molecular Formula Tl
Molecular Weight (g/mol) 204.38
MDL Number MFCD00134063
InChI Key BKVIYDNLLOSFOA-UHFFFAOYSA-N
Synonym ramor, thallium, metallic, thallium, elemental, unii-ad84r52xlf, thallium, soluble compounds, ad84r52xlf, thallium, elemental and soluble compounds, rod, 12.7mm 0.5in dia, granules, thalliumShow MoreShow Less
PubChem CID 5359464
ChEBI CHEBI:30440
IUPAC Name thallium
SMILES [Tl]

Specifications

Quantity 75 mm
Percent Purity 99.999%
Odor Odorless
Weight ≈225g/150mm
Form Rod
Assay (metals basis)
Chemical Name or Material Thallium rod, 12.7mm (0.5 in.) dia.

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberB25T029 Certificate TypeCertificate of Analysis Date17/11/2021 Catalog Number012714.0Y,012714.DO
Lot NumberF16R027 Certificate TypeCertificate of Analysis Date17/11/2021 Catalog Number012714.0Y,012714.DO
Lot NumberB20U045 Certificate TypeCertificate of Analysis Date17/11/2021 Catalog Number012714.0Y,012714.DO
Lot NumberB05T023 Certificate TypeCertificate of Analysis Date17/11/2021 Catalog Number012714.0Y,012714.DO
Lot NumberD12S020 Certificate TypeCertificate of Analysis Date17/11/2021 Catalog Number012714.0Y,012714.DO

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thallium rod, 12.7mm (0.5in) dia, 99.999% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.