Learn More
Julabo™ Thermal Bath Fluids
Used for temperature control applications. Julabo™ Thermal Bath Fluids offer safe and reliable operation.
Brand:  Julabo™ 8940101
 View more versions of this product
Product Code. 11900228
  • 319.00 € / 5 L
Estimated Shipment: 04-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For closed systems like the Presto or Forte
  • Has wide temperature ranges
  • Low viscosity
  • High stability
  • Good heat conductivity
  • Minimum odor
  • Low corrosion tendency
  • Low toxicity
  • Long shelf life
Specifications
Specifications
Transparent
Thermal Bath Fluid
Ideal for temperature control applications and guarantee safe and reliable operation, viscosity, oxi
Thermal M
>255°C
40°C to 170°C
Thermal Bath
Drain tap
5 L
298mm2/s at 20°C