Learn More
Eppendorf™ Thermal printer DPU-S445
Thermal line dot printer with high printing speed and clear type face production.
Brand: Eppendorf™ 6135012007
This item is not returnable.
View return policy
Description
- Automatic print-out of results
- Easy and direct connection via USB
- Compact and lightweight
- Easy paper orientation
- Compatible with the BioPhotometer D30 and the BioSpectrometer product line
Specifications
BioPhotometer D30 and the BioSpectrometer product line | |
Thermal Dot Printer | |
Max printing speed: 90 mm/sec, Dots/line: 832, Resolution: 8 dots/mm | |
GB Plug |
Product Suggestions
Customers who viewed this item also viewed.
Product Content Correction
Product Title
Eppendorf™ Thermal printer DPU-S445 > GB Plug
Spot an opportunity for improvement?Share a Content Correction