Learn More
Brady™ Thermal Transfer Printable Labels
Our B-494 polyester label is a reliable sample identification and traceability solution.
Brand:  Brady™ 306985
 View more versions of this product
Product Code. 17110511
  • 237.00 € / Pack of 3000
Estimated Shipment: 19-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • The label will stay attached and remain legible, even when exposed to chemicals or stored in liquid nitrogen
  • Thanks to subsurface printing, the label's colour code will not fade, smear or smudge and will continue to offer fast sample recognition long after it was printed
Specifications
Specifications
THT-304-494-3-PK
Pink, White
Roll
Front
Labels
BBP™11 Label Printer; BBP™12 Label Printer; Brady IP™ Series Label Printer; BradyPrinter i5100 Industrial Label Printer; BradyPrinter i7100 Industrial Label Printer; THT 76 mm Core Printers
3000 Labels/Roll
-196°C to +70°C
12.70 mm
9.53 mm
25.40 mm
9.53 mm
Acrylic
Gloss
Polyester
Thermal Transfer
Non-sterile
-196°C
-320.8°F to +158°F
0.0762 mm
0.375 in.
1 in.
9.53 mm