missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Thermal Transfer Printable Labels

Our B-494 polyester label is a reliable sample identification and traceability solution.

Brand:  Brady™ 306977

 View more versions of this product

Product Code. 17130501

  • 284.00 € / Pack of 2500
Estimated Shipment: 23-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • The label will stay attached and remain legible, even when exposed to chemicals or stored in liquid nitrogen
  • Thanks to subsurface printing, the label's colour code will not fade, smear or smudge and will continue to offer fast sample recognition long after it was printed
Specifications

Specifications

BPT-310-494-2.5-YL
Yellow, White
Roll
Front
Labels
BBP™11 Label Printer; BBP™12 Label Printer; THT 76 mm Core Printers
2500 Labels/Roll
-196°C to +70°C
15.24 mm
12.70 mm
25.40 mm
Acrylic
Gloss
Polyester
Ink Jet
Non-sterile
-196°C
-320.8°F to +158°F
0.0762 mm
0.5 in.
1 in.
12.70 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.