Learn More
Brady™ Thermal Transfer Printable Labels, B-427
Ideal for a variety of applications. Brady™ Thermal Transfer Printable Labels are made of self-laminating vinyl
Brand:  Brady™ 030541
 View more versions of this product
Product Code. 11578274
  • 463.00 € / 1 roll
Estimated Shipment: 28-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Available in various sizes
Specifications
Specifications
UL Recognized
Thermal Transfer Printable Labels
White, Translucent
Roll
Thermal Transfer Printer Label
Brady 1244, Brady 1344, Brady 2461, Brady 3481, Brady 6441, Brady 300MVP Plus, Brady 200MVP Plus, Brady 300X-Plus II, Brady 360X-Plus II, Brady 600X-Plus II, Tagus T200, Tagus T300, Thermal Transfer Printers, IP300, IP600, BBP81
1500/Pk.
57.15 x 50.08 mm (W x H)
0.004 in.
55.55 mm
2 in.
50.80 mm
50.8 mm
THT-67-427-1.5
Acrylic
Matte
Vinyl, Self-laminating
Cable and wire marking, Panel labeling
1
Rectangle
-70°C to +70°C
0.102 mm
57.15 x 50.80 mm
57.15 mm
2.25 in.
19.05 mm