missing translation for 'onlineSavingsMsg'
Learn More

Texwipe™ ThermaSeal™ 60 Wipers

Feature laser cut, sealed polyester laundered in an ISO class 4 and are ideal for all purpose applications in critical cleanrooms and higher environments

Brand:  Texwipe™ TX2068B

Product Code. 15722396

  • 36.15 € / Pack of 200
Estimated Shipment: 10-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Thermally sealed (laser) edges for particle and fiber control
  • 100% continuous-filament, double-knit polyester fabric
  • Good absorbency, wiping efficiency and abrasion resistance
  • Chemically compatible with standard solvents and alcohols
  • Laundered and packaged in an ISO Class 4 cleanroom environment
  • Solvent-safe Bag-Within-A-Bag™ cleanroom packaging to protect product
  • Complies with REACH and RoHS requirements
  • Color-coded packaging for easy recognition
  • Semiconductor and microelectronics — general wiping and cleaning needs
  • Sterile Applications —Autoclavable
  • USP 797 environments
  • Medical device product cleaning
  • Video display and instrument panel cleaning
Specifications

Specifications

18 x 20 cm
200 Wipers/ Bag, bulk packed, 10 Bags/ Cs.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers