Learn More
Thermographic Measurements Thermax 10 Level Temperature Indicator
Ten level temperature Indicator Strips used to check maximum temperature.Single use, irreversible threshold indicator reports the highest temperature reached
Brand:  Thermographic Measurements M10STHERGAD01P1
 View more versions of this product
Product Code. 17806088
  • 19.80 € / Pack of 10
Edge
Explore more special offers
Description
Description
  • Immediate Response
  • Permanent Record of Highest Temperature Achieved
  • Oil, Water, and Steam Resistant
  • Inexpensive and Easy to Use
  • Self-adhesive
Specifications
Specifications
ISO9001
Laboratory Research, Medical services, Instrumentation, Electronics Industry, Automotive, Aerospace, Steel, Metallurgy, Plastics Industry, Food Industry
51 mm
18 mm
Thermax 10 Level Temperature indicator Range A
71°C
2°C
40°C to 71°C
Electrical Equipment, Pumps, Braking systems, Water Pumps, Aeronautical Equpment, Military Equipment
40, 42, 44, 46, 49, 54, 60, 62, 65, 71°C
18 mm
Single-use
2 in.
40 to 60%
Polyester Film
160°F
36°F
104°F to 160°F
Yes
±1°C
Visual Indicator
°C and °F