Learn More
Thermographic Measurements Thermax 5 Level Temperature Indicator
Five level temperature Indicator Strips used to check maximum temperature. Single use, irreversible threshold indicator reports the highest temperature reached.
Brand:  Thermographic Measurements M05STHERGAD02P1
Product Code. 17808758
  • 14.00 € / Pack of 10
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Immediate Response
  • Permanent Record of Highest Temperature Achieved
  • Oil, Water, and Steam Resistant
  • Inexpensive and Easy to Use
  • Self-adhesive
Specifications
Show More
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermographic Measurements Thermax 5 Level Temperature Indicator > Temperature Range (Metric): 37°C to 46°C