Learn More
Thermo Scientific™ Thermo-Flask™ Benchtop Liquid Nitrogen Containers
Maximize coolant retention with these benchtop liquid nitrogen containers, which contain inner vessels made of borosilicate.
507.53 € - 754.76 €
Product Code | Brand | Capacity (Metric) | Diameter (Metric) | Description | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Diameter (Metric) | Description | Price | Quantity & Availability | |||
11880933
|
Thermo Scientific™
2122 |
1 L | 116 mm | With carrying handle and vented secure lid |
512.55 €
Each |
In Stock
Log in to see stock. |
|||
11868003
|
Thermo Scientific™
2123 |
2 L | 142 mm | With carrying handle and vented secure lid |
550.74 €
Each |
In Stock
Log in to see stock. |
|||
11878003
|
Thermo Scientific™
2124 |
4.5 L | 182 mm | With carrying handle and vented secure lid |
754.76 €
Each |
In Stock
Log in to see stock. |
|||
13488119
|
Thermo Scientific™
2129 |
1 L | 160 mm | Without lid and handle |
534.66 €
Each |
In Stock
Log in to see stock. |
|||
13478129
|
Thermo Scientific™
2130 |
1.9 L | 186 mm | Without lid and handle |
507.53 €
Each |
In Stock
Log in to see stock. |
|||
Description
Maximize coolant retention with light-weight Thermo Scientific™ Thermo-Flask™ Benchtop Liquid Nitrogen Containers, which contain inner vessels of borosilicate and are perfect for everyday cryogenic storage use.
Stainless Steel:- Durable, corrosion-resistant stainless-steel exterior
- Carrying handle and vented secure lid (Part numbers 2122, 2123, 2124)
Shallow Wide Mouth:
- Ideal for use as a cold trap
- Convenient dish-shaped design perfect for benchtop use with crushed ice and dry ice/alcohol
- Stainless-steel jacket and cushioned base for years of trouble-free use
- Without lid and handle (Part number 2129)
- Warranty varies by country, please contact us for details
Specifications
Stainless Steel | |
Benchtop |