Learn More
Thermo Scientific™ 0.2 mL Individual Tubes
Descripción
0.2 mL Individual Tubes with integral “snap shut” caps are suitable for 0.2 mL thermal cycler blocks. They have a 0.25 mL maximum volume when closed. These individual tubes feature thin-wall design and are made from virgin polypropylene. The tubes are cleanroom produced, ensuring that they are free from contaminants that could affect experimental results. They are available in multiple colors with flat or domed caps.
The Thermo Scientific 0.2 mL Individual Tubes with integral ”snap shut“ caps are suitable for 0.2 mL thermal cycler blocks. They have a 0.25 mL maximum volume when closed. These individual tubes feature thin-wall design and are made from virgin polypropylene. The tubes are cleanroom produced, ensuring that they are free from contaminants that could affect experimental results. They are available in multiple colors with flat or domed caps.
Highlights
• Suitable for 0.2 mL thermal cycler blocks
• 0.25 mL maximum tube capacity when closed
•
Integral 'snap shut' cap
• Available with flat or domed caps
Applications
• PCR
• Sample storage
Hazardous:No
>Storage Condition:Ambient
PCR; Sample storage
Certifications
Certified free from human DNA, RNases, and DNases
Especificaciones
Especificaciones
| Certifications/Compliance | RNase, Dnase and human DNA free |
| Color | Yellow |
| Volume (Metric) | 0.2 mL |
| Quantity | 1000 Case |
| For Use With (Equipment) | 0.2mL thermal cycler blocks for PCR and sample storage applications |
| Material | Polypropylene |
| Closure Type | Domed Cap |
| Format | Tube |
| Product Type | Thin Wall PCR Tube |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.