missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ PCR Plate, 96-well, robotic

Product Code. 10445543
Click to view available options
Quantity:
50 plates
Unit Size:
Pack of 50
This item is not returnable. View return policy

Product Code. 10445543

Brand: Thermo Scientific™ AB1300

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

96-well full-skirted SuperPlate which provides 4x more rigidity for superior robotic handling in PCR and qPCR applications.

The Thermo Scientific 96-Well Full-Skirted Plate, Robotic's signature full skirt surround provides an even surface for robotic arms to grip. The SuperPlate design is four times stronger than standard polypropylene plates due to an improved design and a specially selected polymer that minimize warping during thermal cycling and after heat sealing. The plates have thin walls to reduce evaporation and a cut corner at the H1 location. Use these 96-Well Full-Skirted Plates in PCR and qPCR applications.
Highlights
• ANSI:SBS footprint and stackable for use in automated systems
• Low profile to reduce dead space and increase PCR efficiency
• Raised rim design around each well enables secure sealing and safeguards against evaporation
• Uniform, thin well walls deliver maximum and consistent heat transfer for equally high performance from every sample
• 0.2 mL maximum well volume (when used with adhesive and heat seals)
• Cut Corner: H1
Applications
• PCR
• qPCR/Real-Time PCR

Certifications

Certified free from DNase, RNase, and human genomic DNA.

TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Certifications/Compliance Certified RNase/DNase-free
Packaging 50 Plate
Color Clear
Product Type PCR Microplate
For Use With (Equipment) Real-Time PCR Systems or Thermal Cyclers
Skirt Style Skirted
Material Polypropylene
Volume (Metric) Well 0.2 mL
No. of Wells 96
Volume (Metric) Working 0.2 mL
For Use With (Application) PCR, qPCR, RT-PCR
Quantity 50 plates
Reaction Speed Low profile
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.