Learn More
Thermo Scientific™ Aquasil™ C18 Javelin™ Guard Columns
Offer convenient, easy-to-use protection for analytical HPLC columns
Brand:  Thermo Scientific™ 77505-022106
 View more versions of this product
Product Code. 11583311
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Direct-connection, finger-tight design for convenience and maximum efficiency
  • Patented design requires no holder, allowing quick and easy replacement
Features of Aquasil C18 Solid Phase
  • Thermo Scientific polar endcapped C18 phase
  • Alternative selectively compared to conventional C18 phases
  • Retention of polar compounds
  • Compatible with 100% aqueous mobile phases
TRUSTED_SUSTAINABILITY
Specifications
Specifications
5 μm
20 mm
2.1 mm
C18