Learn More
Thermo Scientific™ BioBasic™ 4 Javelin™ Guard Columns
Javelin guard columns offer convenient, easy to use protection for analytical HPLC columns
Brand:  Thermo Scientific™ 72305-021006
Product Code. 11533291
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Direct-connection, fingertight design for convenience and maximum efficiency
  • Patented design requires no holder, allowing quick and easy replacement
  • Based on 300Å silica for outstanding biomolecule performance
  • Lower carbon load for optimal retention of larger peptides and proteins
  • Outstanding reproducibility, efficiency and column lifetime
  • Excellent for LC/MS separations
TRUSTED_SUSTAINABILITY
Specifications
Specifications
5 μm
1 mm
BioBasic
20 mm
BioBasic 4