Learn More
Thermo Scientific™ Capit-All Flex Automated Decapper
The Thermo Scientific™ Capit-All™ Flex Automated Decapper is a full-service, automated capping and decapping solution for 48- and 96-tube racks.
Brand:  Thermo Scientific™ 4120-FLEX
Product Code. 30178456
  • 80245.00 € / Each
Explore more special offers
Description
Description
Experience unparalleled efficiency with our advanced cryotube decapper system, designed to meet the needs of modern laboratories. Compatible with both Matrix and Nunc cryotubes in 96 and 48 formats, this all-in-one instrument combines the functionality of four separate instruments, significantly reducing your laboratory's footprint and costs. Switch seamlessly between tube types without the hassle of adapters or part changes. The large 7 in. angled touchscreen, operable with or without gloves, ensures ease of use and visibility. A built-in drip tray acts as a barrier between caps and tubes to prevent cross-contamination and maintain the integrity of your samples. Our instrument includes tube verification to minimize errors and supports both full and partial racks, adapting to your specific needs. Whether used on the benchtop or an integrated system, it offers flexibility for your storage applications.
  • Compatible with both Matrix and Nunc cryotubes in 96 and 48 formats
  • Functionality of 4 instruments in one, reducing footprint and spend
  • Switch between tube types without the need of adapters or changing parts
  • Large angled 7 in. touchscreen that can be used with or without gloves
  • Built-in drip tray that acts as a barrier between caps and tubes to help prevent cross-contamination and maintain sample integrity
  • Tube verification to prevent errors
  • Works with full or partial racks of tubes
  • Can be used for benchtop or integrated applications - fits inside a Biosafety Cabinet
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Matrix and Nunc tubes in 96 and 48 format
Automated Decapper
100/240 VAC
96-Channel
Touchscreen User Interface
Ω
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products