missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Easy Pierce Heat Sealing Foil

Use this heat sealing foil to ready your microplate for compound management and high-throughput screening applications, such as PCR.

Brand:  Thermo Scientific™ AB-1797

571.59 EUR valid until 2024-12-31
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 30067381

  • 783.00 € / Pack of 1000

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Seal your microplate for compound management and high-throughput screening, including PCR, with Thermo Scientific™ Easy Pierce Heat Sealing Foil. Suitable for use with Dimethyl sulfoxide (DMSO) reagents, this sealing foil creates a peelable bond that can be easily pierced and removed by hand.

Features:

  • Quick operation
  • Safe for temperature-sensitive samples
  • Designed for reliable performance with Thermo Scientific ALPS instruments, including the, ALPS 3000, ALPS 25 and ALPS 50 V
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Clear
Clear Seal Thin, Polymer 80x125
Non-sterile
96
1000
Ideal for fluorescence, colorimetric, and optical applications; suitable for PCR/ qPCR; ELISA
ALPS 3000, ALPS 25 and ALPS 50 V; AB-1443A
Abgene
No
-80°C to +120°C
Polypropylene, Polyethylene, Polystyrene
Yes
Life Science Reagent Resistant
Polyester
Not Autoclavable
1000
80 x 125 mm
General; Long term
Heat Seal
Polypropylene
Yes
All well shapes
General
Low Autofluorescence
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.