Learn More
Thermo Scientific™ Evolution™ One, OnePlus and Pro Spectrophotometer Solid Sample Holders
Designed to hold solid samples for transmittance measurements
Brand:  Thermo Scientific™ 222-217400
 View more versions of this product
Product Code. 10061104
  • 975.00 € / Each
Explore more special offers
Description
Description
Designed to hold solid samples for transmittance measurements
Solid Sample Holder
  • Provides easy mounting of solid samples like filters, plastic, glass and optics
  • Accommodates standard 2 x 3 in. sample holders
Universal Sample Holder
  • Holds round or square optical samples; glass or plastic samples; lenses from sunglasses or goggles
  • Requires one Solid Sample Holder (14-387-503)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Universal Sample Holder
Thermo Scientific™ Evolution™ One, OnePlus and Pro UV-visible spectrophotometers
Round or square optical samples, glass or Plastic samples, lenses from sunglasses or goggles