missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ ATP Solution, Tris buffered

Thermo Scientific ATP (adenosine 5'-triphosphate), Tris buffered is an extremely stable nucleotide, supplied as 100 mM aqueous solution titrated to pH 7.3-7.5 with Tris base.

Brand:  Thermo Scientific™ R1441

34.65 EUR valid until 2025-06-30
Use promo code "24074" to get your promotional price.


Product Code. 15235116

  • 46.06 € / 0.25mL

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific ATP (adenosine 5'-triphosphate), Tris buffered is an extremely stable nucleotide, supplied as 100 mM aqueous solution titrated to pH 7.3-7.5 with Tris base. The nucleotide has greater than 99% purity and is free of nuclease activities and functionally tested in in vitro transcription.

Features
• Purity confirmed by HPLC
• Free of trace contaminating nucleotides, endo-, exodeoxyribonuclease and ribonuclease activities
• Higher in vitro transcription yields with Tris buffered NTPs when compared to NTPs titrated with NaOH
• Stable for 3 years at -20°C, and after multiple freeze-thaw cycles

Applications
in vitro transcription
• RNA amplification
• siRNA synthesis
• aRNA synthesis
• Ligation
• Phosphorylation

Related products
• GTP, Tris Buffered (Cat. No. R1461)
• UTP, Tris Buffered (R1471)
• NTP Set, Tris Buffered (R1481)

TRUSTED_SUSTAINABILITY
Specifications

Specifications

ATP Solution, Tris buffered, 100 mM (0.25 mL)

Store at –20°C.
100 mM
0.25 mL
Liquid
Dry Ice
Unlabeled
in vitro Transcription, RNA Amplification
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2901775 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberR1441
Lot Number2980224 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog NumberR1441
Lot Number2946601 Certificate TypeCertificate of Analysis Date26/01/2025 Catalog NumberR1441
Lot Number3008693 Certificate TypeCertificate of Analysis Date26/01/2025 Catalog NumberR1441
Lot Number3023389 Certificate TypeCertificate of Analysis Date26/01/2025 Catalog NumberR1441
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ ATP Solution, Tris buffered > 0.25mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.