missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ NTP Set, Tris buffered

Thermo Scientific NTP Set, Tris buffered is a convenient set of 100 mM ATP, CTP, GTP, and UTP solutions.

Brand:  Thermo Scientific™ R1481

Product Code. 15285116

  • 138.00 € / 100000nmol

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific NTP Set, Tris buffered is a convenient set of 100 mM ATP, CTP, GTP, and UTP solutions. These extremely stable nucleotides are supplied as aqueous solutions titrated to pH 7.3-7.5 with Tris base. The nucleotides have greater than 99% purity and are free of nuclease activities and functionally tested in in vitro transcription.

Features
• Purity confirmed by HPLC
• Free of trace contaminating nucleotides, endo-, exodeoxyribonuclease and ribonuclease activities
• Higher in vitro transcription yields with Tris buffered NTPs when compared to NTPs titrated with NaOH
• Stable for 3 years at -20°C, and after multiple freeze-thaw cycles

Applications
in vitro transcription
• RNA amplification
• siRNA synthesis
• aRNA synthesis

Related products
• ATP, Tris Buffered (Cat. No. R1441)
• GTP, Tris Buffered (R1461)
• UTP, Tris Buffered (R1471)

TRUSTED_SUSTAINABILITY
Specifications

Specifications

NTP Set, Tris buffered, 100 mM (4 x 0.25 mL)

Store at –20°C.
100 mM
4 x 0.25 mL
Liquid
Dry Ice
Unlabeled
in vitro Transcription, RNA Amplification
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.