missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TopVision Agarose Tablets

High quality, multipurpose agarose tablets in blister packaging.

149.00 € - 552.00 €

Specifications

Product Type Top Vision Agarose Tablet
Separation Range 100 to >30000 bp
Melting Point 88 °C
Product Line TopVision
Validated Application Gel Electrophoresis
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15225116
View Documents
Thermo Scientific™
R2801
200 Tablets (0.5 g each)
149.00 €
200 tablets
In Stock
to see stock.
Edge
Add to basket
 
15200143
View Documents
Thermo Scientific™
R2802
1000 Tablets (0.5 g each)
552.00 €
1000 tablets
In Stock
to see stock.
Add to basket
 
Description

Description

Description
TopVision Agarose Tablets, low EEO (LE), is multipurpose agarose in tablet format delivered in a convenient blister pack. The agarose tablets are made of standard melting point agarose that yields high resolution, sharp DNA bands with high clarity and low background. Its optimized gel strength enhances ease of gel processing and handling. Manufactured using an innovative Organic Solvent-Free manufacturing process that is greener and more environmentally friendly.

Features

  • Convenient tablet format - no messy weighing required.
  • Fast dissolving protocol.
  • “Green”- manufactured with significantly reduced environmental impact, using an innovative organic solvent-free technology.
  • Enhanced resolution and reduced background.
Applications
Ideal for routine DNA and RNA gel electrophoresis and blotting assays.

Environmentally responsible Green Solution
Unlike traditional agarose powder, which production relies heavily on the use of toxic organic solvents, TopVision Agarose Tablets are manufactured using an innovative process that completely eliminates dependency on organic solvents and significantly reduces environmental impact while creating a safer working environment.

Appearance:White to off-white powder
> Ash:≤ 0.5%
> CAS:9012-36-6
> EEO:≤ 0.13
> Gel Strength:≥ 1200 g/cm2 (1% Gel)
> Gelling Point:36°C ± 1.5°C (1.5% gel)
> Hazardous:No
> Impurities (DNAses, RNases and Proteases):None
> Melting Point:88°C ± 1.5°C (1.5% gel)
> Moisture:≤ 10%
> Shelf Life:24 Months
> Shipping Condition:Ambient
> Solubility:Clear colorless solution at 1g in 100 mL water
> Storage Condition:Ambient
> Sulfate:≤ 0.15%

Specifications

Specifications

Top Vision Agarose Tablet
88 °C
Gel Electrophoresis
100 to >30000 bp
TopVision
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 2 Years

SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.