Learn More
Thermo Scientific™ GeneJET™ Plasmid Maxiprep Kit Reagents
Reagents for use with the GeneJET Plasmid Maxiprep Kit. The Plasmid Maxiprep Kit is designed for large scale isolation of high quality plasmid DNA from recombinant E. coli cultures.
Specifications
Content And Storage | Store at room temperature |
---|---|
For Use With (Equipment) | GeneJET™ Endo-free Plasmid Maxiprep Kit |
Description
Reagents for use with the Thermo Scientific GeneJET Plasmid Maxiprep Kit (K0861). The GeneJET Plasmid Maxiprep Kit utilizes silica-based membrane technology in the form of convenient spin column. Each prep recovers up to 750 μg of high copy plasmid DNA that can be used in a wide variety of molecular biology procedures, such as restriction endonuclease digestion, PCR, cloning, transformation, automated sequencing, in vitro transcription, and transfection of robust cell lines.
Highlights
- Efficient - high yields up to 750 μg of high quality plasmid DNA from 250 mL of bacterial culture
- High concentration of DNA purified - > 400 ng/μL
- Convenient - spin column format, centrifuge or vacuum protocols
- Fast - procedure takes 60 minutes, considerably faster than using gravity flow columns
- Isolated plasmid DNA is suitable for all conventional molecular biology procedures, including:
- Transfection of robust cell lines
- FastDigest restriction enzyme digestion
- Automated fluorescent sequencing
- PCR
- Cloning
- Transformation
- In vitro transcription
Hazardous: No
Quality Control:
- The GeneJET Plasmid Maxiprep Kit is qualified by isolating high copy number plasmid DNA from 250 mL of overnight E. coli culture (OD600 = 2) grown in LB medium following the protocol outlined in the manual.
- The kit passes QC requirements if the purified plasmid DNA has an A260/280 ratio between 1.8 and 1.9 and a dominant band of supercoiled plasmid DNA is observed after agarose gel electrophoresis.
- The functional quality of the plasmid DNA is evaluated by digestion with restriction endonucleases.
Isolated plasmid DNA is suitable for all conventional molecular biology procedures, including:
- Transfection of robust cell lines
- FastDigest restriction enzyme digestion
- Automated fluorescent sequencing
- PCR
- Cloning
- Transformation
- In vitro transcription
Order Info
Orders placed Monday through Wednesday ship next day; Thursday and Friday orders ship the following Monday.
Specifications
Store at room temperature | |
GeneJET™ Endo-free Plasmid Maxiprep Kit |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.