missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Hypercarb™ Drop-in Guard Cartridges

Offer convenience, economy and effective protection for extending column lifetime.

Brand:  Thermo Scientific™ 35007-014001

Product Code. 11538310

  • 479.90 € / Pack of 4
Estimated Shipment: 07-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • The 10mm design offers maximum protection with minimal increase in retention
  • Compatible with Thermo Scientific Uniguard™ direct connection and stand-alone holders
Features of Hypercarb Solid Phase
  • Suited to solve problem separations
  • Exceptional retention of very polar analytes
  • True orthoganality to C18
  • Separate structurally related substances
  • Excellent for high temperature applications

Order Info

Drop-in cartridges are available in other Thermo Scientific phases. Please contact your Customer Service Representative for more information

TRUSTED_SUSTAINABILITY
Specifications

Specifications

7 μm
4.6 mm
Hypercarb
10 mm
Hypercarb
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.