missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Hypercarb™ Javelin™ Guard HPLC Columns

Offer convenient, easy-to-use protection for analytical HPLC columns

510.00 € - 639.43 €

Specifications

Particle Size 5 μm
Stationary Phase Hypercarb
View More Specs

Products 2
Product Code Brand Diameter (Metric) Inner Length (Metric) Price Quantity & Availability  
Product Code Brand Diameter (Metric) Inner Length (Metric) Price Quantity & Availability  
11573201
View Documents
Thermo Scientific™
35005-012106
2.1 mm 10 mm
639.43 €
Pack of 2
Estimated Shipment: 22-01-2025
to see stock.
Add to basket
 
11543211
View Documents
Thermo Scientific™
35005-024006
4 mm 20 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Direct-connection, fingertight design for convenience and maximum efficiency
  • Patented design requires no holder, allowing quick and easy replacement
  • Suited to solve problem separations
  • Exceptional retention of very polar analytes
  • True orthoganality to C18
  • Separate structurally related substances
  • Excellent for high temperature applications

Specifications

Specifications

5 μm
Hypercarb
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.