Learn More
Thermo Scientific™ Hypersil™ ODS (C18) Javelin™ Guard Columns
Offer convenient, easy-to-use protection for analytical HPLC columns
Brand:  Thermo Scientific™ 30105-014006
 View more versions of this product
Product Code. 11503181
  • 441.49 € / Pack of 4
Estimated Shipment: 10-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Direct-connection, fingertight design for convenience and maximum efficiency
  • Patented design requires no holder, allowing quick and easy replacement
TRUSTED_SUSTAINABILITY
Specifications
Specifications
5 μm
10 mm
4 mm
C18