Learn More
Thermo Scientific™ HyPURITY™ C8 Guard Cartridges
Drop-in guard cartridges offer convenience, economy and effective protection for extending column lifetime
Brand:  Thermo Scientific™ 22205-012101
Product Code. 10714437
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Requires : Thermo Scientific UNIGUARD direct connection or stand alone holder, sold separately.
Description
Description
  • The 10mm design offers maximum protection with minimal increase in retention
  • Fit Thermo Scientific UNIGUARD™ direct connection and stand alone holders
  • High quality, highly pure silica columns
TRUSTED_SUSTAINABILITY
Specifications
Specifications
5 μm
10 mm
2.1 mm
C8