Learn More
Unity Lab Services™ Thermo Scientific™ Laboratory Refrigerators & Freezers (+5 to -40°C) Validation Documentation for RDE, TDE, FDE and IUE Series
As an OEM service provider, all protocols designed to optimize efficiency are included with each Thermo Scientific Laboratory Refrigerators and Freezers (+5C to -40C) Validation Documention for All Thermo Scientific laboratory refrigerators and freezers, except TSX505 and Jewett fridge/freezer
Brand:  Unity Lab Services™ IOQDOCE89003482
Product Code. 12787853
  • 1200.00 € / Each
Explore more special offers
Description
Description
IQ/OQ documentation that supports regulatory requirements and industry standards for instrument compliance. Our documents are kept up to date by conducting annual reviews. We work directly with our product experts to have the most updated documentation available.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Thermo Scientific Laboratory Refrigerators and Freezers (+5 to -40°C) including all Thermo Scientific laboratory refrigerators and freezers, except TSX505 and Jewett fridge/freezer
Documentation Only for the following, Services Not Included: Validation (IQ/OQ) includes a standardized protocol procedure, IQ: Verify Installation and environmental conditions, OQ: Verify operation to design specification, open door and power loss testing.
Contact your local sales representative for a list of standard and extended coverage areas.
IQ/OQ Documentation