Learn More
Thermo Scientific™ Carboy Kit
For Use With: Large capacity reach-in CO2 Incubator
Brand:  Thermo Scientific™ 191596
Product Code. 11881130
  • 513.56 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
For Use With: Large capacity reach-in CO2 Incubator
TRUSTED_SUSTAINABILITY
Specifications
Specifications
7.8 L
Large capacity reach-in CO2 incubator
LED
Carboy Kit
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Carboy Kit > Carboy Kit; Includes: autoclavable 2 gallon (7.8 liters) carboy; valve; adapter; hose; and mounting bracket