Learn More
Thermo Scientific™ Low Protein Binding Microcentrifuge Tubes
Minimize protein sample loss during sample manipulation, especially when there are extensive processing steps, by using Thermo Scientific Pierce Low Protein Binding Microcentrifuge Tubes.
Brand:  Thermo Scientific™ 90411
228.95 EUR valid until 2024-12-31
Use promo code "21938" to get your promotional price.
 View more versions of this product
Product Code. 15352617
  • 241.00 € / Pack of 2500
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Thermo Scientific Pierce Low Protein Binding Microcentrifuge Tubes (1.5 or 2 mL) help minimize protein sample loss during sample manipulation and are ideal in applications where processing steps are extensive.
Pierce Low Protein Binding Microcentrifuge Tubes help minimize sample loss in mass spectrometry and immunoprecipitation/co-immunoprecipitation applications. The tubes are conveniently packaged at 250 tubes per package to minimize contamination even when purchased in bulk.
Durable—use at speeds up to 20,000 RCF
Compatible—suitable for IP and mass spectrometry applications
Nuclease-free—certified RNase/DNase-free polypropylene tubes
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Collection Tube
2500 x 1.5 mL Tubes
Pierce™
Low Protein Binding Tube
1.5 mL