missing translation for 'onlineSavingsMsg'
Learn More

Nunc™ Thermo Scientific™ Nunc™ Non-Coded Cryobank Vial Systems
GREENER_CHOICE

Store diagnostic and biological samples down to vapor-phase liquid nitrogen in Thermo Scientific™ Nunc™ Non-Coded Cryobank Vial Systems

Brand:  Nunc™ 374223-BR

639.00 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.


Product Code. 15533714

  • 990.00 € / Pack of 480
Estimated Shipment: 17-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Store diagnostic and biological samples down to vapor-phase liquid nitrogen in Thermo Scientific™ Nunc™ Non-Coded Cryobank Vial Systems

Store diagnostic and biological samples down to vapor-phase liquid nitrogen in Thermo Scientific™ Nunc™ Non-Coded Cryobank Vial Systems, compatible with capping/decapping equipment and labeling options. Cryobank products are available in a volume range of 0.5L through 5.0L, and complimented by a range of certifications. 

Secure Tracking
  • Solid one-piece construction eliminates the risk of labels falling off during cryogenic storage, preserving the identity of valuable sample material

Superior Storage Format
  • Cryobank tubes are available in space-conscious microplate format racks or can be stored in traditional cryoboxes, accomodating your preferred storage format
  • Microplate format is compatible with your existing multichannel pipettes and automated liquid handling equipment
  • Unique anti-rotation feature allows compatibility with automated capping/decapping equipment in single, 8-channel, or full rack options to accomodate multiple throughput requirements
  • TPE gasketed polypropylene cap provides superior seal during high or low temperature storage
  • Low binding surface allows maximum recovery of low concentration samples

Certified to meet your storage needs
  • CE certified and conforms to IATA requirements for diagnostic use and transportation
  • Certified DNAse/RNAse free (0.5/1.0mL), non-pyrongenic (LAL-test), non-toxic (USP class VI test)
  • Sterility assurance level 10-6 for the most sensitive sample storage

Stringent Quality Control
  • Leak testing validation performed during the manufacturing process to ensure sample integrity is maintained
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Clear
2 mL
48
None
Racked, 48
Sterile
480
Nunc
Polypropylene
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers