missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ COD (Chemical Oxygen Demand) Calibration Standards for AQUAfast Colorimeters

Product Code. p-2362929
Click to view available options
Concentration:
1000 ppm
10000 ppm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10316522

Brand: Thermo Scientific™ CODS01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Use these calibration standards to check your colorimeter instrument operations before measuring COD chemistries, for example in environmental and wastewater monitoring.

Use Thermo Scientific™ COD (Chemical Oxygen Demand) Calibration Standards with Orion™ AQUAfast Colorimeters such as AQ4000 Advanced Colorimeter and Orion AQUAfast II AQ2040 Colorimeter. Orion colorimeters, which measure COD chemistries, are ideal for environmental and wastewater monitoring. Use these calibration standards to check operations prior to using the instruments.

  • For use with: Orion AQUAfast AQ4000, Orion AQUAfast AQ2040
  • Two Calibration Standards: 1000 ppm and 10000ppm, both in 475 mL bottles

Order Info

Reagents sold separately.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1000 ppm
Certifications/Compliance EPA
For Use With (Equipment) AQUAfast Colorimeter
Industry Type Environmental Testing, Wastewater Monitoring
Quantity 475 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.