Learn More
Thermo Scientific™ Orion™ Plastic-Membrane ISE Sensor Modules
Brand:  Thermo Scientific™ 931801
Product Code. 10749741
  • 356.78 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Interchangeable sensor modules for Orion Plastic-Membrane electrodes. Combination sensors can be used in combination elctrtodes; half-cell sensors in half-cell models. Orion Electrode Body/Handle is available separately: order 13-641-24.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Ammonium ISE
Measures ammonium in aqueous solutions quickly, simply, accurately and economically
32°F to 122°F
Sensor Module
Built in ATCC
0°C to 50°C