missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Owl™ ADJ1 Adjustable System Accessories and Replacement Parts
Extremely compact at only 16cm wide
17.80 € - 65.20 €
Product Code | Brand | Type | Dimensions (L x W) | Thickness (English) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Dimensions (L x W) | Thickness (English) | Price | Quantity & Availability | |||
11590295
|
Thermo Scientific™
ADJ1-40G |
Blank Glass Plate | 15.7 x 6.2 in. (40 x 16 cm) | 0.18 in. |
26.00 €
Each |
Estimated Shipment: 29-04-2025
Log in to see stock. |
|||
11550295
|
Thermo Scientific™
ADJ1-23G |
Offset Glass Plate | 9.05 x 6.2 in. (23 x 16 cm) | 0.18 in. |
20.00 €
Each |
Estimated Shipment: 29-04-2025
Log in to see stock. |
|||
11500305
|
Thermo Scientific™
ADJ1-40LM |
Heat Sink | 15.7 x 6.2 in. (40 x 16 cm) | 0.18 in. |
65.20 €
Pack of 2 |
Estimated Shipment: 29-04-2025
Log in to see stock. |
|||
11580295
|
Thermo Scientific™
ADJ1-38G |
Offset Glass Plate | 14.9 x 6.2 in. (38 x 16 cm) | 0.18 in. |
26.00 €
Each |
Estimated Shipment: 29-04-2025
Log in to see stock. |
|||
11510305
|
Thermo Scientific™
ADJ1-40R |
Notched Glass Plate | 15.7 x 6.2 in. (40 x 16 cm) | 0.18 in. |
62.10 €
Each |
Estimated Shipment: 29-04-2025
Log in to see stock. |
|||
11520305
|
Thermo Scientific™
ADJ1-C25 |
Spacer Set | 9.8 x 0.39 in. (25 x 1 cm) | 0.03 in. |
17.90 €
Each |
Estimated Shipment: 29-04-2025
Log in to see stock. |
|||
11560305
|
Thermo Scientific™
ADJ1-GK |
Gasket |
18.00 €
Each |
Estimated Shipment: 29-04-2025
Log in to see stock. |
|||||
11540305
|
Thermo Scientific™
ADJ1-D25 |
Spacer Set | 9.8 x 0.39 in. (25 x 1 cm) | 0.05 in. |
17.80 €
Each |
Estimated Shipment: 29-04-2025
Log in to see stock. |
|||
Description
- The height of the upper buffer chamber (UBC) may be adjusted by sliding the UBC up or down two support poles allowing gels to vary in height from 14 cm to 40 cm.
- The notched or offset glass plate of the gel sandwich presses against a silicone gasket in the UBC for a leakproof seal.
- Plates are clamped to the UBC with either binder clamps or heavy duty pony clamps.
Specifications
Owl ADJ1 Adjustable Electrophoresis System |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title