missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Phusion Hot Start II High-Fidelity PCR Master Mixes

Thermo Scientific Phusion High-Fidelity DNA polymerases set a gold standard for high performance PCR.

Brand:  Thermo Scientific™ F565L

566.80 EUR valid until 2024-12-31
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 15311732

  • 872.00 € / 500 reactions

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: Phusion Hot Start II High–Fidelity DNA Polymerase
  • Phusion Hot Start II High–Fidelity DNA Polymerase (2U/μL)
  • 5x Phusion HF Buffer
  • 5x Phusion GC Buffer
  • DMSO
  • 50mM MgCl2 solution
Phusion Green Hot Start II High–Fidelity DNA Polymerase
  • Phusion Hot Start II High–Fidelity DNA Polymerase (2U/Glc muL)
  • 5x Phusion Green HF Buffer
  • 5x Phusion Green GC Buffer
  • DMSO
  • 50mM MgCl2 solution
Both Phusion and Phusion Green Buffer provide 1.5 mM MgCl2 in the final 1x concentration
This item is not returnable. View return policy

Description

Description

Thermo Scientific Phusion High-Fidelity DNA Polymerases set a gold standard for high performance PCR. Featuring an error rate 50-fold lower than that of Taq, and 6-fold lower than that of Pfu, Phusion High-Fidelity DNA Polymerase is excellent choice for cloning and other applications requiring high fidelity. Phusion DNA Polymerases offer robust performance with short protocol times, even in the presence of PCR inhibitors, and generate higher yields with lower enzyme amounts than other DNA polymerase.

Highlights

  • Reaction set up at room temperature
  • No non-specific amplification and primer degradation during reaction set up
  • Zero-time reactivation due to unique hot start technology
  • High fidelity (52X Taq)
  • Fast PCR due to short extension times (15-30 s/kb)
  • Robust reactions, minimal optimization needed
  • Increased product yields with minimal enzyme amounts
  • Direct loading on gels

With Phusion Hot Start II High-Fidelity DNA Polymerases amplification proceeds without the production of nonspecific products due to the combination of Phusion DNA Polymerase and a reversibly bound, specific Affibody ligand that inhibits DNA polymerase activity at room temperature. The Affibody ligand also inhibits the 3'→5' exonuclease activity of the polymerase, thus preventing degradation of primers and template DNA during reaction set up. At temperatures that promote polymerase activity, the ligand is released, rendering the polymerase fully active. Phusion Hot Start II DNA Polymerase is immediately reactivated at high temperatures so it does not require a separate activation step in PCR protocols.

Phusion Green Hot Start II High-Fidelity PCR Master Mix is convenient 2X mix designed to minimize the number of pipetting steps. The master mix contains Phusion Hot Start II DNA Polymerase, nucleotides and optimized reaction buffer including MgCl2. The buffer also includes a density reagent and two tracking dyes for direct loading of PCR products on a gel.

Applications

  • High-fidelity PCR
  • High throughput
  • Amplification of difficult (GC-rich) templates
  • Template generation for sequencing
  • Multiplex PCR
  • Long-range PCR
  • Cloning
  • Mutagenesis
  • Microarray

Using Phusion DNA Polymerases

Annealing rules for Phusion DNA Polymerases are different from many common DNA polymerases (such as Taq DNA polymerases).

TRUSTED_SUSTAINABILITY
Specifications

Specifications

• 2X Phusion Hot Start II High-Fidelity PCR Master Mix
• Nuclease-free water
• DMSO

2X Phusion Hot Start II High-Fidelity PCR Master Mix provides 1.5 mM MgCl2 in the final 1X concentration.
Store at –20°C.
Phusion Hot Start II High-Fidelity DNA Polymerase
Hot Start High-Fidelity PCR Master Mix
Dry Ice
2X
Built-In Hot Start
Blunt
20 kb or less
Medium
Fast
500 x 50 μL Reactions
Hot-start PCR, High-fidelity PCR
52X
500 Reactions
SuperMix or Master Mix
Colorless
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.