missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Phusion Hot Start II High-Fidelity PCR Master Mixes

Product Code. 15341732 Shop All Thermo Scientific Products
Click to view available options
Color:
Colorless
Green
No. of Reactions:
100 Reactions
500 Reactions
Unit Size:
100 reactions
500 reactions
This item is not returnable. View return policy

Product Code. 15341732

Brand: Thermo Scientific™ F566L

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Includes

Phusion Hot Start II High–Fidelity DNA Polymerase

  • Phusion Hot Start II High–Fidelity DNA Polymerase (2U/μL)
  • 5x Phusion HF Buffer
  • 5x Phusion GC Buffer
  • DMSO
  • 50mM MgCl2 solution
Phusion Green Hot Start II High–Fidelity DNA Polymerase
  • Phusion Hot Start II High–Fidelity DNA Polymerase (2U/Glc muL)
  • 5x Phusion Green HF Buffer
  • 5x Phusion Green GC Buffer
  • DMSO
  • 50mM MgCl2 solution
Both Phusion and Phusion Green Buffer provide 1.5 mM MgCl2 in the final 1x concentration

Thermo Scientific Phusion High-Fidelity DNA polymerases set a gold standard for high performance PCR.

Thermo Scientific Phusion High-Fidelity DNA Polymerases set a gold standard for high performance PCR. Featuring an error rate 50-fold lower than that of Taq, and 6-fold lower than that of Pfu, Phusion High-Fidelity DNA Polymerase is excellent choice for cloning and other applications requiring high fidelity. Phusion DNA Polymerases offer robust performance with short protocol times, even in the presence of PCR inhibitors, and generate higher yields with lower enzyme amounts than other DNA polymerase.

Highlights

  • Reaction set up at room temperature
  • No non-specific amplification and primer degradation during reaction set up
  • Zero-time reactivation due to unique hot start technology
  • High fidelity (52X Taq)
  • Fast PCR due to short extension times (15-30 s/kb)
  • Robust reactions, minimal optimization needed
  • Increased product yields with minimal enzyme amounts
  • Direct loading on gels

With Phusion Hot Start II High-Fidelity DNA Polymerases amplification proceeds without the production of nonspecific products due to the combination of Phusion DNA Polymerase and a reversibly bound, specific Affibody ligand that inhibits DNA polymerase activity at room temperature. The Affibody ligand also inhibits the 3'→5' exonuclease activity of the polymerase, thus preventing degradation of primers and template DNA during reaction set up. At temperatures that promote polymerase activity, the ligand is released, rendering the polymerase fully active. Phusion Hot Start II DNA Polymerase is immediately reactivated at high temperatures so it does not require a separate activation step in PCR protocols.

Phusion Green Hot Start II High-Fidelity PCR Master Mix is convenient 2X mix designed to minimize the number of pipetting steps. The master mix contains Phusion Hot Start II DNA Polymerase, nucleotides and optimized reaction buffer including MgCl2. The buffer also includes a density reagent and two tracking dyes for direct loading of PCR products on a gel.

Applications

  • High-fidelity PCR
  • High throughput
  • Amplification of difficult (GC-rich) templates
  • Template generation for sequencing
  • Multiplex PCR
  • Long-range PCR
  • Cloning
  • Mutagenesis
  • Microarray

Using Phusion DNA Polymerases

Annealing rules for Phusion DNA Polymerases are different from many common DNA polymerases (such as Taq DNA polymerases).

TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X
Content And Storage • 2X Phusion Green Hot Start II High-Fidelity PCR Master Mix (provides 1.5 mM MgCl2 in the final 1X concentration)
• Nuclease-free water
• DMSO

Store at -20°C.
GC-Rich PCR Performance Medium
Polymerase Phusion Hot Start II High-Fidelity DNA Polymerase
Reaction Speed Fast
Product Type Hot Start High-Fidelity PCR Master Mix
Quantity 500 reactions
Shipping Condition Dry Ice
For Use With (Application) Hot-start PCR, High-fidelity PCR
Fidelity (vs. Taq) 52X
Hot Start Built-In Hot Start
No. of Reactions 500 Reactions
Overhang Blunt
Reaction Format SuperMix or Master Mix
Size (Final Product) 20 kb or less
Color Green
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.