missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Phusion U Multiplex PCR Master Mixes

Product Code. 15341722 Shop All Thermo Scientific Products
Click to view available options
Color:
Colorless
Green
No. of Reactions:
100 Reactions
500 Reactions
Unit Size:
100 reactions
500 reactions
This item is not returnable. View return policy

Product Code. 15341722

Brand: Thermo Scientific™ F562S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thermo Scientific Phusion U Multiplex PCR Master Mix is a ready-to-use, 2X end-point PCR master mix designed for simultaneous amplification of multiple targets in a single tube.

Thermo Scientific Phusion U Green Multiplex PCR Master Mix is a ready-to-use, 2X end-point PCR master mix designed for simultaneous amplification of multiple targets in a single tube. Over 20 primer pairs may be combined into a single reaction for highly specific and efficient multiplexing over a broad range of primer and template concentrations.

The Phusion U Multiplex PCR Master Mix contains a proprietary buffer with balanced concentrations of all PCR components eliminating the need for tedious optimization. The Green format further simplifies the workflow - it includes a density reagent and two electrophoresis tracking dyes for direct loading of PCR products on gels.

Highlights

  • Specific and efficient multiplex PCR of over 20 targets up to 2.5 kb
  • Excellent results with templates of suboptimal purity due to inhibitor tolerance
  • Fast cycling and direct loading of PCR products on gels

Applications

  • Genotyping
  • Pathogen detection
  • Food testing
  • Analysis of genetically modified organisms
  • Amplification of microsatellites

The master mix is based on Phusion U Hot Start DNA Polymerase, a high performance enzyme developed using fusion technology. Similarly to other enzymes from Phusion family, Phusion U DNA Polymerase incorporates more nucleotides per binding event than any other DNA polymerase. This allows achieving high yields of PCR products with shorter extention times and consequently reduced total protocol times. Significantly enhanced processivity also enables successful multiplex PCR on difficult targets such as GC-rich templates or templates of suboptimal purity. Being highly tolerant of many PCR inhibitors, Phusion U Multiplex PCR Master Mixes can even be used with unpurified samples such as blood or serum.

Specificity of multiplex PCR is increased by Affibody-mediated hot start mechanism. Reversibly bound Affibody ligand inhibits the polymerase at ambient temperatures preventing amplification of nonspecific products and formation of primer-dimers.

Using Phusion DNA Polymerases

Annealing rules for Phusion DNA Polymerases are different from many common DNA polymerases (such as Taq DNA polymerases).

TRUSTED_SUSTAINABILITY

Spezifikation

Concentration 2X
Content And Storage • Phusion U Hot Start DNA Polymerase
• 2X Reaction buffer
• 3 mM MgCl2
• 400 μM of each dATP, dTTP, dCTP and dGTP
• Nuclease-free water

Store at –20°C.
GC-Rich PCR Performance Medium
Polymerase Phusion U Hot Start DNA Polymerase
Reaction Speed Fast
Product Type Multiplex PCR Master Mix
Quantity 100 reactions
Shipping Condition Dry Ice
For Use With (Application) Hot-start PCR, Genotyping
Fidelity (vs. Taq) 25X
Hot Start Built-In Hot Start
No. of Reactions 100 Reactions
Overhang Blunt
Reaction Format SuperMix or Master Mix
Size (Final Product) 2.5 kb or less
Color Colorless
Mehr anzeigen Weniger anzeigen

For Research Use Only. Not for use in diagnostic procedures.

Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt