missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Phusion U Multiplex PCR Master Mixes

Product Code. 15351722 Shop All Thermo Scientific Products
Click to view available options
Color:
Colorless
Green
No. of Reactions:
100 Reactions
500 Reactions
Unit Size:
100 reactions
500 reactions
This item is not returnable. View return policy

Product Code. 15351722

Brand: Thermo Scientific™ F562L

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thermo Scientific Phusion U Multiplex PCR Master Mix is a ready-to-use, 2X end-point PCR master mix designed for simultaneous amplification of multiple targets in a single tube.

Thermo Scientific Phusion U Green Multiplex PCR Master Mix is a ready-to-use, 2X end-point PCR master mix designed for simultaneous amplification of multiple targets in a single tube. Over 20 primer pairs may be combined into a single reaction for highly specific and efficient multiplexing over a broad range of primer and template concentrations.

The Phusion U Multiplex PCR Master Mix contains a proprietary buffer with balanced concentrations of all PCR components eliminating the need for tedious optimization. The Green format further simplifies the workflow - it includes a density reagent and two electrophoresis tracking dyes for direct loading of PCR products on gels.

Highlights

  • Specific and efficient multiplex PCR of over 20 targets up to 2.5 kb
  • Excellent results with templates of suboptimal purity due to inhibitor tolerance
  • Fast cycling and direct loading of PCR products on gels

Applications

  • Genotyping
  • Pathogen detection
  • Food testing
  • Analysis of genetically modified organisms
  • Amplification of microsatellites

The master mix is based on Phusion U Hot Start DNA Polymerase, a high performance enzyme developed using fusion technology. Similarly to other enzymes from Phusion family, Phusion U DNA Polymerase incorporates more nucleotides per binding event than any other DNA polymerase. This allows achieving high yields of PCR products with shorter extention times and consequently reduced total protocol times. Significantly enhanced processivity also enables successful multiplex PCR on difficult targets such as GC-rich templates or templates of suboptimal purity. Being highly tolerant of many PCR inhibitors, Phusion U Multiplex PCR Master Mixes can even be used with unpurified samples such as blood or serum.

Specificity of multiplex PCR is increased by Affibody-mediated hot start mechanism. Reversibly bound Affibody ligand inhibits the polymerase at ambient temperatures preventing amplification of nonspecific products and formation of primer-dimers.

Using Phusion DNA Polymerases

Annealing rules for Phusion DNA Polymerases are different from many common DNA polymerases (such as Taq DNA polymerases).

TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X
Content And Storage • Phusion U Hot Start DNA Polymerase
• 2X Reaction buffer
• 3 mM MgCl2
• 400 μM of each dATP, dTTP, dCTP and dGTP
• Nuclease-free water

Store at –20°C.
GC-Rich PCR Performance Medium
Polymerase Phusion U Hot Start DNA Polymerase
Reaction Speed Fast
Product Type Multiplex PCR Master Mix
Quantity 500 reactions
Shipping Condition Dry Ice
For Use With (Application) Hot-start PCR, Genotyping
Fidelity (vs. Taq) 25X
Hot Start Built-In Hot Start
No. of Reactions 500 Reactions
Overhang Blunt
Reaction Format SuperMix or Master Mix
Size (Final Product) 2.5 kb or less
Color Colorless
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.