missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Anti-HA Agarose

HA Epitope Tag Monoclonal antibody 26181 and 26182 from Thermo Scientific™ specifically detects HA Epitope Tag in Tag samples, and it is validated for Immunoprecipitation

Brand:  Thermo Scientific™ 26181


 View more versions of this product

Product Code. 11533060

  • 708.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

HA Epitope Tag Monoclonal antibody 26181 and 26182 from Thermo Scientific™ specifically detects HA Epitope Tag in Tag samples, and it is validated for Immunoprecipitation

An HA tag is frequently incorporated into recombinant proteins for a variety of purposes. An anti-HA antibody can then be used to detect the protein when doing studies with transfected cells.
Host Species: Mouse
Cross Reactivity: Tag
Isotype: IgG1
Clone: 2-2.2.14
Immunogen: HA peptide YPYDVPDYA derivitized to ovalbumin
Formulation: 1mL Purified Antibody in PBS, with 0.05% sodium azide; pH7.2
Synonyms: human influenza hemagglutinin, HA-tag, HA, HA Epitope Tag
26181 is supplied as 1mL of antibody on crosslinked 4% beaded agarose supplied as a 50% slurry in 0.1M phosphate, 0.15M NaCl, pH 7.2 with 0.05% sodium azide. Specific for the HA peptide YPYDVPDYA. The anti-HA agarose has been used successfully in immunoprecipitation applications. The anti-HA agarose has 3.5mg mouse anti-HA IgG1 monoclonal antibody conjugated permL of settled agarose resin. HA Synthetic Peptide (Product # 26184) is available for use in neutralization and control experiments and to competitively elute HA-tagged fusion proteins from immobilized anti-HA affinity resin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Agarose
Bottle
Anti-HA Antibody
1 mL
Agarose Resin, Affinity
HA-epitope tag (YPYDVPDYA)
1 mL of settle resin, provided as 2 mL of 50% slurry. Store at 2–8°C.
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Pierce Anti-HA Agarose

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Supplemental information
  • MIXTURE LIST-Contains : isopropanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.