missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Firefly Luc One-Step Glow Assay Kit

Product Code. p-4701929 Shop All Thermo Scientific Products
Click to view available options
Quantity:
100 Reaction Kit
1000 Reaction Kit
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 15275663

Brand: Thermo Scientific™ 16197

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Perform firefly luciferase assays in high-throughput screening applications.

The Thermo Scientific Pierce Firefly One-Step Luc Glow Assay Kit measures firefly luciferase activity in mammalian cell culture using a single reagent-addition step, making it ideal for high-throughput screening (HTS) applications.

This glow-type luciferase assay kit is designed for use with cultured mammalian cells that are transfected with vectors expressing the appropriate firefly luciferase reporter enzyme. Several features make the kit especially suitable for luminometers without injectors and for high-throughput screening (HTS) applications. The convenient, one-step, homogenous protocol minimizes handling steps to support the use of automation. Simply combine the supplied substrate solution and assay buffer to make a single working solution, and then add it to microplate wells containing the transfected, treated cells. The stable signal output (glow) of this system provides flexibility with regard to incubation time. When used with Thermo Scientific Firefly Luc Plasmids, the kit provides a highly sensitive bioluminescent reporter assay system for the detection of promoter or pathway activity.

Highlights:

  • Sensitive - highly sensitive detection of firefly luciferase activity
  • Convenient - one-step, mix-and-read protocol; no additional reagents required
  • Stable - increased signal half-life and stability compared to flash assays
  • Simple - does not require a luminometer with injectors
  • Automation-friendly - amenable to high throughput assays
Includes:
Assay buffer and substrate solution (100X)
Requires:
Firefly luciferase and luminometer or other instrument capable of monitoring luminescence, such as Thermo Scientific Luminoskan Ascent and Varioskan Flash Microplate Readers.
Applications:
  • Promoter studies for analyzing cis- and trans-acting regulatory elements
  • Drug screening
  • siRNA and miRNA screening
  • Multiplexed assays to study off-target effects
  • Protein localization reporter assays
  • Signal transduction pathway analysis
Product Details:
Firefly luciferase is a 62kDa protein produced in nature by several species of the Lampyridae family of beetles, which includes the genera Photinus and Luciola. Thermo Scientific Red Firefly Luciferase has an emission range of 560-700nm (λmax = 613nm). Bioluminescent signal from firefly luciferase originates from the oxidation of D-Luciferin. Light output captured using a luminometer can be correlated with the amount of firefly luciferase protein produced and used to determine the activity of the promoter driving firefly luciferase expression. Firefly luciferase bioluminescence provides researchers with a non-radioactive assay for measuring transcriptional activity of regulatory elements. Firefly luciferase reporter activity can be used to determine promoter activity, to study upstream pathway activity in the cell, or to determine effect of activators or inhibitors.

TRUSTED_SUSTAINABILITY

Specifications

Type Assay Kit
Target Luciferase, Firefly Luciferase, Cypridina Luciferase
Sufficient For 1000 Luciferase Assays in Microplate Wells
Quantity 1000 Reaction Kit
Content And Storage • Firefly Luc One-Step Substrate, Lyophilized (makes 1 mL of 1X), store at -20°C
• Firefly Luc One-Step Assay Buffer, 115 mL, store at -20°C

Upon receipt store kit at -20°C.
Assay Reporter Enzyme, Luciferase Reporter Assay
Compatible Cells Mammalian Cells
Detection Method Bioluminescence
For Use With (Equipment) Luminometer (Microplate)
Format 384-well plate, 96-well plate
Label Type Enzyme Labeled
Substrate Properties Chemical Substrate
Technique Enhanced Chemiluminescence
Substrate Type Luciferase Substrate
Product Line Pierce
Substrate D-Luciferin
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.