Learn More
Thermo Scientific™ Neuron Isolation Enzyme (with papain) for Pierce™ Primary Cell Isolation Kits
Easily isolate and culture highly viable functional primary neurons
Brand: Thermo Scientific™ 88285
Description
The Thermo Scientific Pierce Primary Neuron Isolation Kit provides isolation and culturing reagents for the optimal yield and viability of primary neurons form embryonic cortical and hippocampal tissue from mouse and rat.
The Primary Neuron Isolation Kit has been designed to obtain high viability and purity of primary embryonic neurons using validated reagents and an optimized procedure for enzymatic digestion and culturing. The kit contains tissue-specific dissociation enzymes, media formulated specifically for neuronal cell culture, and cell culture supplements that promote the isolation and growth of functional neurons. The validated protocol and reagents produce highly pure primary neurons for either short- or long-term culture, and the kit is designed for use by both experienced and non-experienced users.
Highlights:
- Optimized - procedure and reagents optimized for viability, yield, purity, and ease-of-use
- Yield - provides a 2-fold increase in yield compared to do-it-yourself methods
- Viability - greater than 95% of isolated cells remain viable in culture from Day 1 to Week 4
- Functional - cultured neurons contain processes indicative of complete differentiation and functionality
- Complete kit includes Neuronal Isolation Enzyme, Hanks' Balanced Salt Solution (HBSS), Neuronal Culture Medium, Neuronal Culture Media Supplement, Glutamine Supplement, and Neuronal Growth Supplement
- Fresh embryonic mouse or rat tissue sample, heat-inactivated fetal bovine serum (FBS), poly-D-lysine treated culture dishes
- Neuronal cell differentiation
- Immunohistochemistry (IHC)
- Functional and biochemical assays
- Biologically relevant system for preclinical drug discovery, neurotoxicity testing, and predictive disease modeling
The Pierce Primary Neuron Isolation Kit has been optimized to provide excellent cell yield and to sustain cells at greater than 95% viability. The neurons are appropriately polarized, develop extensive axonal and dendritic arbors, express neuronal and synaptic markers, and form numerous, functional synaptic connections with one another. They can be used as a model system for molecular and cellular biology studies of neuronal development and function, especially for visualizing the subcellular localization of endogenous or expressed proteins, imaging protein, neuronal polarity, and dendritic growth and synapse formation [1].
General References:
1. Silva, R.F.M, et al. (2006) Dissociated primary nerve cell cultures as models for assessment of neurotoxicity. Toxicology Letters 163:1-9.
2. Ivenshitz, M. and Segal, M, (2009). Neuronal density determines network connectivity and spontaneous activity in cultured hippocampus. J Neurophysiol 104:1052-60.
Specifications
Cell Isolation Enzyme | |
Neuronal Cells (Pre-Natal), Primary Cells | |
Solid | |
1 Vial |
Stability: 1 year at -20°C Storage of Reconstituted Enzyme: -20°C for 6 months and up to two freeze-thaw cycles, or 1 week at 4°C. Enzyme use after prolonged storage may result in poor performance. |
|
Rat, Mouse | |
Pierce™ |
Safety and Handling
- Neuronal Isolation Enzyme (with papain)
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Papain
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.