Learn More
Thermo Scientific™ Reacti-Vial Reaction Vials
An internal cone makes small sample handling easy and convenient
Brand:  Thermo Scientific™ TS-13223
205.11 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 11752257
  • 349.00 € / Pack of 12
Estimated Shipment: 18-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Extra thick glass wall magnifies the sample, making these vials ideal for observing chemical reactions
  • Amber vials available for light-sensitive compounds
  • Supplied complete with Open-Top screw caps and PTFE/Rubber discs
Residue isolation, Derivative preparation, Maximum sample retrieval, Moisture protection, Sample storage, Precipitations, Centrifugations, Solvent evaporation
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Clear
5 mL
5 mL
Certificate of Analysis (COA)
Silica Glass
Internal
Screw Cap
Vial