missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ RevertAid RT Reverse Transcription Kit

The Thermo Scientific RevertAid RT Kit is a complete system for efficient synthesis of first strand cDNA from mRNA or total RNA templates.

Brand:  Thermo Scientific™ K1691

Product Code. 15255146

  • 974.00 € / 500 reactions

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

The Thermo Scientific RevertAid RT Kit is a complete system for efficient synthesis of first strand cDNA from mRNA or total RNA templates. The kit uses RevertAid Reverse Transcriptase (RT), which has lower RNase H activity compared to AMV reverse transcriptase. The enzyme maintains activity at 42–50°C and is suitable for synthesis of cDNA up to 13 kb. The recombinant RiboLock RNase Inhibitor, supplied with the kit, effectively protects RNA from degradation at temperatures up to 55°C.

First strand cDNA synthesized with this system can be directly used as a template in PCR or real-time PCR. It is also ideal as a first step for second strand cDNA synthesis or linear RNA amplification. Radioactively and non-radioactively labeled nucleotides can be incorporated into first strand cDNA for use as a probe in hybridization experiments, including microarrays.

Features of the RevertAid RT Kit include:
• Full-length first strand cDNA up to 13 kb
• Optimum reaction temperature 42°C
• Complete kit includes all the components for RT reactions

Applications
• First strand cDNA synthesis for RT-PCR and RT-qPCR
• Construction of full length cDNA libraries
• Antisense RNA synthesis

Order Info

Orders placed Monday through Wednesday ship next day; Thursday and Friday orders ship the following Monday.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

• RevertAid Reverse Transcriptase
• Ribolock RNase Inhibitor
• 5X Reaction Buffer
• dNTP Mix
• Random Hexamer Primer
• Nuclease-free water

Store at –20°C.


60 min.
42°C
RevertAid
Dry Ice
First-Strand cDNA
Separate components
Up to 13 kb
Kit
Reverse Transcription
500 reactions
Yes
Real Time PCR (qPCR)
500 Reactions
Reverse Transcription
RNA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.