Learn More
Thermo Scientific™ RockIt Rocking bioreactor BioProcess Container (BPC)
Brand:  Thermo Scientific SH30980.03
 View more versions of this product
Product Code. 17958151
  • 710.00 € / Pack of 5
Explore more special offers
Specifications
Bioprocess Container for Liquid
Single-Use Bioreactor
Sterile
Thermo Scientific
Gamma Irradiated
5 cm (2 in)
5 cm (2 in)
C-flex
C-flex
P.C. Silicone
22.25 ft.
26.12 ft.
20 L
Plastic Film
5 Panel ports
1
100 cm (39.4 in)
105 cm (41.3 in)
10 cm (3.9 in)
P.C. Silicone
P.C. Silicone
22.25 x 26.12 ft
678.18 cm
796.14 cm