Learn More
Thermo Scientific™ Rotors for Savant™ DNA SpeedVac™ Concentrators
Accessory Rotors
961.79 € - 1256.25 €
Specifications
Type | Glass And Plastic Tube |
---|---|
For Use With (Equipment) | Savant™ DNA SpeedVac™ concentrator rotor |
Product Code | Brand | Type | Holds | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Holds | Price | Quantity & Availability | ||||
11728539
|
Thermo Scientific™
RH20-12 |
Glass And Plastic Tube | 20 Tubes |
961.79 €
Each |
Estimated Shipment: 17-02-2025
Log in to see stock. |
||||
11718579
|
Thermo Scientific™
RH6-25 |
Glass And Plastic Tube | 6 x 30 mL Tubes |
1256.25 €
Each |
Estimated Shipment: 17-02-2025
Log in to see stock. |
||||
Description
Easy to use circular rotors with different tube hole sizes to accommodate different sample sizes. These rotors require no additional accessories and fit directly into your concentrator.
Specifications
Glass And Plastic Tube | |
Savant™ DNA SpeedVac™ concentrator rotor |