missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ ROX Solution, 50 μM

ROX dye is added to the qPCR reactions to serve as an internal reference for normalization of the fluorescent signal when using real-time instruments that can detect ROX.

Brand:  Thermo Scientific™ R1371

Product Code. 13273189

  • 111.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific ROX dye is added to the qPCR reactions to serve as an internal reference for normalization of the fluorescent signal when using real-time instruments that can detect ROX. ROX allows for the correction of well-to-well variation appearing due to pipetting inaccuracies and fluorescence fluctuations. It can be added to an entire 2X master mix tube or to an individual reaction mixture (for recommended amounts of ROX Solution for a specific real-time instrument refer to the qPCR manual). ROX does not participate in PCR and does not interfere with the specificity or sensitivity of qPCR assays.
ROX Solution contains ROX dye as a 50μM aqueous solution at pH 7.6. The excitation maximum and emission maximum of ROX are at 580nm and 621nm, respectively.

Thermo Scientific ROX dye is added to the qPCR reactions to serve as an internal reference for normalization of the fluorescent signal when using real-time instruments that can detect ROX. ROX allows for the correction of well-to-well variation appearing due to pipetting inaccuracies and fluorescence fluctuations. It can be added to an entire 2X master mix tube or to an individual reaction mixture (for recommended amounts of ROX Solution for a specific real-time instrument refer to the qPCR manual). ROX does not participate in PCR and does not interfere with the specificity or sensitivity of qPCR assays.
ROX Solution contains ROX dye as a 50 μM aqueous solution at pH 7.6. The excitation maximum and emission maximum of ROX are at 580 nm and 621 nm, respectively.

Hazardous:No
>Spectral properties:ROX dye excitation/emission maxima are at 580 nm/621 nm, respectively.
> Storage Condition:-20°C
>-20°C
> Storage note:Note that ROX dye is light sensitive. Exposure to light should be minimized.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Contains:
• 1 mL ROX Solution, 50 μM

Store at -20°C.
Tube
qPCR
ROX
1 mL
2 X
Primer-probe
Low
ROX
qPCR Reagents
50 μM
1 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ ROX Solution, 50 μM > 1mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.