Learn More
Epredia™ Cassette 1 Biopsy Cassettes
Specially designed to hold biopsy specimens during processing and embedding
Brand: Epredia™ 1001090
This item is not returnable.
View return policy
Description
- Made of acetal polymer
- Cassette writing face is at a 30° angle
- Pore style is small square
- Locking lid attached by two hinges
- Supplied in a box of 500 cassettes with attached breakaway lids
Order Info
Not available in Tube Packs
Specifications
Square | |
Hinged | |
Acetal Polymer |
30° | |
Pink | |
Biopsy Cassette |