Learn More
Epredia™ Shandon™ Plastic Coverplates
For use on Shandon Sequenza™ and Cadenza™ Immunostainers
164.00 € - 969.00 €
Specifications
Product Type Coverplate
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11927774
View Documents
Epredia™
72110013
250/Cs.
969.00 €
Pack of 250
12766807
View Documents
Epredia™
72110017
25/Pk.
164.00 €
Pack of 25
Description
Description
Saves on the use of costly reagents.
  • Device that fits over standard microscope slide allowing specimens to be immunostained with a minimum amount of reagent
  • Frees considerable technician time
  • Eliminates handling of slides between steps
  • Allows sections to be placed virtually anywhere on slide
  • Protects specimen from drying during long incubations
  • Ensures consistency in staining quality
Specifications
Specifications
Coverplate
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Epredia™ Shandon™ Plastic Coverplates