missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Shandon™ Pathcentre™ Tissue Processor Accessories

For use with the Epredia™ Shandon™ Tissue Processor

22.30 € - 1040.00 €

Specifications

For Use With (Equipment) Tissue Processor
View More Specs

Products 2
Product Code Brand Product Type Quantity Price Quantity & Availability  
Product Code Brand Product Type Quantity Price Quantity & Availability  
12775108
View Documents
Epredia™
A75210193
Storage Bottle 3 Pk. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12808220
View Documents
Epredia™
66110048
Airbag 4 Pk. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Specifications

Tissue Processor
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.